6-Amino-5-chloronicotinonitrile ≥95%
Supplier: Matrix Scientific
Synonyms:
2-Amino-3-chloro-5-cyanopyridine
042630-500MG
042630-1G
042630-5G
101856-690EA
120
USD
101856-690
101856-692
101856-694
6-Amino-5-chloronicotinonitrile ≥95%
6-Amino-5-chloronicotinonitrile
Hazard: Irritant
Formula:
C₆H₄ClN₃ MW: 153.5691 g/mol Melting Pt: 185…188 °C |
MDL Number:
MFCD03617768 CAS Number: 156361-02-3 |
Specification Test Results
Melting point | 185-188°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...