2-(4-Fluorophenyl)-1-hydrazinecarbothioamide ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(4-Fluorophenyl)thiosemicarbazide

042626-500MG 042626-1G 042626-5G
101856-670EA 278.46 USD
101856-670 101856-672 101856-674
2-(4-Fluorophenyl)-1-hydrazinecarbothioamide ≥95%
2-(4-Fluorophenyl)-1-hydrazinecarbothioamide

Hazard: Irritant

Formula: C₇H₈FN₃S
MW: 185.22 g/mol
Melting Pt: 189…191 °C
MDL Number: MFCD01566642
CAS Number: 280133-47-3

Order Now

Specification Test Results

Melting point 189-191°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR