3-[3-Methoxy-4-(2-propynyloxy)phenyl]acrylic acid ≥95%
Supplier: Matrix Scientific
042622-500MG
042622-1G
042622-5G
101856-654EA
400
USD
101856-654
101856-656
101856-658
3-[3-Methoxy-4-(2-propynyloxy)phenyl]acrylic acid ≥95%
3-[3-Methoxy-4-(2-propynyloxy)phenyl]acrylic acid
Hazard: Irritant
Formula:
C₁₃H₁₂O₄ Melting Pt: 205…207 °C |
MDL Number:
MFCD02379952 CAS Number: 385383-50-6 |
Specification Test Results
Melting point | 205-207°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...