[3-Methoxy-4-(2-propynyloxy)phenyl]methanol ≥95%

Supplier: Matrix Scientific

042621-500MG 042621-1G 042621-5G
101856-648EA 400 USD
101856-648 101856-650 101856-652
[3-Methoxy-4-(2-propynyloxy)phenyl]methanol ≥95%
[3-Methoxy-4-(2-propynyloxy)phenyl]methanol

Hazard: Irritant

Formula: C₁₁H₁₂O₃
MW: 192.22 g/mol
Melting Pt: 58…60 °C
MDL Number: MFCD03001222
CAS Number: 385383-48-2

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR