3-Methoxy-4-(2-propynyloxy)benzenecarbaldehydeoxime ≥95%

Supplier: Matrix Scientific

Synonyms: (E)-N-{[3-Methoxy-4-(prop-2-yn-1-yloxy)phenyl]methylidene}hydroxylamine

042620-500MG 042620-1G 042620-5G
101856-642EA 375.38 USD
101856-642 101856-644 101856-646
3-Methoxy-4-(2-propynyloxy)benzenecarbaldehydeoxime ≥95%
3-Methoxy-4-(2-propynyloxy)benzenecarbaldehydeoxime

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MW: 205.22 g/mol
Melting Pt: 120…122 °C
MDL Number: MFCD03001221
CAS Number: 385383-46-0

Order Now

Specification Test Results

Melting point 120-122°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR