3-Amino-3-(4-ethylphenyl)propionic acid ≥95%
Supplier: Matrix Scientific
042617-500MG
042617-1G
042617-5G
101856-626EA
181.54
USD
101856-626
101856-628
101856-630
3-Amino-3-(4-ethylphenyl)propionic acid ≥95%
3-Amino-3-(4-ethylphenyl)propionic acid
Hazard: Irritant
Formula:
C₁₁H₁₅NO₂ Melting Pt: 218-220°C |
MDL Number:
MFCD01990387 CAS Number: 117391-52-3 |
Specification Test Results
Melting point | 218-220°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...