2-[4-Chloro(phenylsulfonyl)anilino]acetic acid ≥95%
Supplier: Matrix Scientific
042615-500MG
042615-1G
042615-5G
101856-616EA
556.92
USD
101856-616
101856-618
101856-620
2-[4-Chloro(phenylsulfonyl)anilino]acetic acid ≥95%
2-[4-Chloro(phenylsulfonyl)anilino]acetic acid
Hazard: Irritant
Formula:
C₁₄H₁₂ClNO₄S Melting Pt: 161…162 °C |
MDL Number:
MFCD00406209 CAS Number: 117309-41-8 |
Specification Test Results
Melting point | 161-162°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...