2-(4-Benzoylphenyl)-2-(4-fluorophenyl)acetonitrile ≥95%
Supplier: Matrix Scientific
042614-500MG
042614-1G
042614-5G
101856-610EA
181.54
USD
101856-610
101856-612
101856-614
2-(4-Benzoylphenyl)-2-(4-fluorophenyl)acetonitrile ≥95%
2-(4-Benzoylphenyl)-2-(4-fluorophenyl)acetonitrile
Hazard: Irritant
Formula:
C₂₁H₁₄FNO MW: 315.35 g/mol Melting Pt: 88…90 °C |
MDL Number:
MFCD01316984 CAS Number: 446275-89-4 |
Specification Test Results
Melting point | 88-90°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...