Methyl-4-isopropyl-1,2,3-thiadiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

042607-500MG 042607-1G 042607-5G
101856-586EA 556.92 USD
101856-586 101856-588 101856-590
Methyl-4-isopropyl-1,2,3-thiadiazole-5-carboxylate ≥95%
Methyl-4-isopropyl-1,2,3-thiadiazole-5-carboxylate

Hazard: Irritant

Formula: C₇H₁₀N₂O₂S
Melting Pt: 120…122 °C
MDL Number: MFCD03001214
CAS Number: 446275-88-3

Order Now

Specification Test Results

Melting point 120-122°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR