2-(4-Acetylphenoxy)-6-chlorobenzenecarbonitrile ≥95%
Supplier: Matrix Scientific
042606-500MG
042606-1G
042606-5G
101856-580EA
375.38
USD
101856-580
101856-582
101856-584
2-(4-Acetylphenoxy)-6-chlorobenzenecarbonitrile ≥95%
2-(4-Acetylphenoxy)-6-chlorobenzenecarbonitrile
Hazard: Irritant
Formula:
C₁₅H₁₀ClNO₂ Melting Pt: 112…114 °C |
MDL Number:
MFCD00105059 CAS Number: 337920-47-5 |
Specification Test Results
Melting point | 112-114°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...