3-(2-Fluoro-4-nitrophenoxy)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042605-500MG 042605-1G 042605-5G
101856-574EA 375.38 USD
101856-574 101856-576 101856-578
3-(2-Fluoro-4-nitrophenoxy)-2-thiophenecarboxylic acid ≥95%
3-(2-Fluoro-4-nitrophenoxy)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₆FNO₅S
Melting Pt: 135…137 °C
MDL Number: MFCD00202749
CAS Number: 337919-93-4

Order Now

Specification Test Results

Melting point 135-137°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR