2-Chloro-6-(3,4-dichlorophenyl)nicotinonitrile ≥95%

Supplier: Matrix Scientific

042603-500MG 042603-1G
101856-564EA 278.46 USD
101856-564 101856-566
2-Chloro-6-(3,4-dichlorophenyl)nicotinonitrile ≥95%
2-Chloro-6-(3,4-dichlorophenyl)nicotinonitrile

Hazard: Irritant

Formula: C₁₂H₅Cl₃N₂
Melting Pt: 163…166 °C
MDL Number: MFCD08056600
CAS Number: 885950-30-1

Order Now

Specification Test Results

Melting point 163-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR