2-Chloro-6-(4-chlorophenyl)nicotinonitrile ≥95%
Supplier: Matrix Scientific
042602-500MG
042602-1G
042602-5G
101856-558EA
375.38
USD
101856-558
101856-560
101856-562
2-Chloro-6-(4-chlorophenyl)nicotinonitrile ≥95%
2-Chloro-6-(4-chlorophenyl)nicotinonitrile
Hazard: Irritant
Formula:
C₁₂H₆Cl₂N₂ Melting Pt: 180…183 °C |
MDL Number:
MFCD00215059 CAS Number: 84596-41-8 |
Specification Test Results
Melting point | 180-183°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...