3-(2-Nitrophenoxy)-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-Nitrophenoxy)thiophene-2-carboxylic acid

042597-500MG 042597-1G 042597-5G
101856-536EA 278.46 USD
101856-536 101856-538 101856-540
3-(2-Nitrophenoxy)-2-thiophenecarboxylic acid ≥95%
3-(2-Nitrophenoxy)-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₇NO₅S
Melting Pt: 165…167 °C
MDL Number: MFCD00202726
CAS Number: 252026-99-6

Order Now

Specification Test Results

Melting point 165-167°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR