3-(4-Nitrophenoxy)-2-thiophenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-(4-Nitrophenoxy)thiophene-2-carboxylic acid
042595-500MG
042595-1G
042595-5G
101856-524EA
278.46
USD
101856-524
101856-526
101856-528
3-(4-Nitrophenoxy)-2-thiophenecarboxylic acid ≥95%
3-(4-Nitrophenoxy)-2-thiophenecarboxylic acid
Hazard: Irritant
Formula:
C₁₁H₇NO₅S Melting Pt: 171…173 °C |
MDL Number:
MFCD00202718 CAS Number: 252026-66-7 |
Specification Test Results
Melting point | 171-173°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...