Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate ≥95%

Supplier: Matrix Scientific

042594-500MG 042594-1G 042594-5G
101856-518EA 278.46 USD
101856-518 101856-520 101856-522
Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate ≥95%
Methyl-3-(2-fluoro-4-nitrophenoxy)-2-thiophenecarboxylate

Hazard: Irritant

Formula: C₁₂H₈FNO₅S
MW: 297.26 g/mol
Melting Pt: 112…114 °C
MDL Number: MFCD00202715
CAS Number: 252026-46-3

Order Now

Specification Test Results

Melting point 112-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR