5-{[4-(Trifluoromethyl)phenyl]sulfanyl}-1,2,3-thiadiazole-4-carboxylic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 5-[4-(Trifluoromethyl)phenylthio]-1,2,3-thiadiazole-4-carboxylic acid

042593-500MG 042593-1G
101856-514EA 723.08 USD
101856-514 101856-516
5-{[4-(Trifluoromethyl)phenyl]sulfanyl}-1,2,3-thiadiazole-4-carboxylic acid ≥95%
5-{[4-(Trifluoromethyl)phenyl]sulfanyl}-1,2,3-thiadiazole-4-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₅F₃N₂O₂S₂
MW: 306.29 g/mol
Melting Pt: 162…166 °C
MDL Number: MFCD01936015
CAS Number: 338982-07-3

Order Now

Specification Test Results

Melting point 162-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR