1,1-Diphenylethylene oxide ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2-Diphenyloxirane

042592-500MG 042592-1G 042592-5G
101856-508EA 278.46 USD
101856-508 101856-510 101856-512
1,1-Diphenylethylene oxide ≥95%
1,1-Diphenylethylene oxide

Hazard: Flammable Solid/Irritant/Stored under Argon/Store in Freezer

Formula: C₁₄H₁₂O
MW: 196.249 g/mol
Melting Pt: 54…56 °C
MDL Number: MFCD00040725
CAS Number: 882-59-7

Order Now

Specification Test Results

Melting point 54-56°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR