1-Methyl-4-(4-nitrobenzoyl)-1H-pyrrole-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

042591-500MG 042591-1G 042591-5G
101856-502EA 278.46 USD
101856-502 101856-504 101856-506
1-Methyl-4-(4-nitrobenzoyl)-1H-pyrrole-2-carbaldehyde ≥95%
1-Methyl-4-(4-nitrobenzoyl)-1H-pyrrole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₀N₂O₄
Melting Pt: 210…212 °C
MDL Number: MFCD00172169
CAS Number: 128843-59-4

Order Now

Specification Test Results

Melting point 210-212°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR