4-Benzoyl-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%

Supplier: Matrix Scientific

042590-500MG 042590-1G 042590-5G
101856-496EA 93.85 USD
101856-496 101856-498 101856-500
4-Benzoyl-1-methyl-1H-pyrrole-2-carbaldehyde ≥95%
4-Benzoyl-1-methyl-1H-pyrrole-2-carbaldehyde

Hazard: Irritant

Formula: C₁₃H₁₁NO₂
Melting Pt: 112…114 °C
MDL Number: MFCD00172168
CAS Number: 128843-58-3

Order Now

Specification Test Results

Melting point 112-114°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR