4-Hydroxy-1-(2-thienylsulfonyl)-2-pyrrolidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042585-500MG 042585-1G 042585-5G
101856-472EA 921.54 USD
101856-472 101856-474 101856-476
4-Hydroxy-1-(2-thienylsulfonyl)-2-pyrrolidinecarboxylic acid ≥95%
4-Hydroxy-1-(2-thienylsulfonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₉H₁₁NO₅S₂
Melting Pt: 204…206 °C
MDL Number: MFCD00202702
CAS Number: 251310-43-7

Order Now

Specification Test Results

Melting point 204-206°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR