Methyl-4-chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-pyrrolidinecarboxylate ≥95%
Supplier: Matrix Scientific
042584-500MG
042584-1G
101856-468EA
1109.23
USD
101856-468
101856-470
Methyl-4-chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-pyrrolidinecarboxylate ≥95%
Methyl-4-chloro-1-[3-chloro-5-(trifluoromethyl)-2-pyridinyl]-2-pyrrolidinecarboxylate
Hazard: Irritant
Formula:
C₁₂H₁₁Cl₂F₃N₂O₂ MW: 343.13 g/mol Melting Pt: 84…87 °C |
MDL Number:
MFCD00664157 CAS Number: 251310-42-6 |
Specification Test Results
Melting point | 84-87°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...