5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carbohydrazide ≥95%

Supplier: Matrix Scientific

042583-500MG 042583-1G
101856-464EA 1483.08 USD
101856-464 101856-466
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carbohydrazide ≥95%
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carbohydrazide

Hazard: Irritant

Formula: C₁₁H₁₁ClF₃N₅O₂
MW: 337.69 g/mol
Melting Pt: 173…175 °C
MDL Number: MFCD00202621
CAS Number: 251310-32-4

Order Now

Specification Test Results

Melting point 173-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR