1-[(3,4-Dimethoxyphenyl)sulfonyl]-4-hydroxy-2-pyrrolidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042582-500MG 042582-1G 042582-5G
101856-458EA 556.92 USD
101856-458 101856-460 101856-462
1-[(3,4-Dimethoxyphenyl)sulfonyl]-4-hydroxy-2-pyrrolidinecarboxylic acid ≥95%
1-[(3,4-Dimethoxyphenyl)sulfonyl]-4-hydroxy-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₇NO₇S
Melting Pt: 169…172 °C
MDL Number: MFCD00202700
CAS Number: 251097-85-5

Order Now

Specification Test Results

Melting point 169-172°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR