5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carboxylic acid ≥95%

Supplier: Matrix Scientific

042581-500MG 042581-1G
101856-454EA 1483.08 USD
101856-454 101856-456
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carboxylic acid ≥95%
5-({[3-Chloro-5-(trifluoromethyl)-2-pyridinyl]amino}methyl)-4,5-dihydro-3-isoxazole carboxylic acid

Hazard: Irritant

Formula: C₁₁H₉ClF₃N₃O₃
MW: 323.66 g/mol
Melting Pt: 161…163 °C
MDL Number: MFCD00202620
CAS Number: 251097-82-2

Order Now

Specification Test Results

Melting point 161-163°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR