2-{[(2-Chlorophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%

Supplier: Matrix Scientific

042579-500MG 042579-1G 042579-5G
101856-442EA 375.38 USD
101856-442 101856-444 101856-446
2-{[(2-Chlorophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%
2-{[(2-Chlorophenyl)sulfonyl]amino}-4-methylpentanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₄S
Melting Pt: 164…166 °C
MDL Number: MFCD00173034
CAS Number: 251097-66-2

Order Now

Specification Test Results

Melting point 164-166°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR