2-{[(4-Bromophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%
Supplier: Matrix Scientific
042577-500MG
042577-1G
042577-5G
101856-430EA
181.54
USD
101856-430
101856-432
101856-434
2-{[(4-Bromophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%
2-{[(4-Bromophenyl)sulfonyl]amino}-4-methylpentanoic acid
Hazard: Irritant
Formula:
C₁₂H₁₆BrNO₄S Melting Pt: 128…130 °C |
MDL Number:
MFCD00172816 CAS Number: 68305-78-2 |
Specification Test Results
Melting point | 128-130°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...