2-{[(4-Fluorophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Fluorobenzenesulphamido)-4-methylpentanoic acid

042576-500MG 042576-1G 042576-5G
101856-424EA 278.46 USD
101856-424 101856-426 101856-428
2-{[(4-Fluorophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%
2-{[(4-Fluorophenyl)sulfonyl]amino}-4-methylpentanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆FNO₄S
MW: 289.33 g/mol
Melting Pt: 124…126 °C
MDL Number: MFCD00172812
CAS Number: 251097-25-3

Order Now

Specification Test Results

Melting point 124-126°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR