2-{[(4-Chlorophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%

Supplier: Matrix Scientific

042575-1G 042575-5G
101856-420EA 350.77 USD
101856-420 101856-422
2-{[(4-Chlorophenyl)sulfonyl]amino}-4-methylpentanoic acid ≥95%
2-{[(4-Chlorophenyl)sulfonyl]amino}-4-methylpentanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆ClNO₄S
Melting Pt: 136…138 °C
MDL Number: MFCD00172811
CAS Number: 68305-77-1

Order Now

Specification Test Results

Melting point 136-138°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR