3-[(4-Nitrobenzyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%
Supplier: Matrix Scientific
042571-500MG
042571-1G
101856-410EA
375.38
USD
101856-410
101856-412
3-[(4-Nitrobenzyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%
3-[(4-Nitrobenzyl)sulfanyl]-2-thiophenecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₉NO₄S₂ MW: 295.34 g/mol Melting Pt: 198…201 °C |
MDL Number:
MFCD01316316 CAS Number: 251097-01-5 |
Specification Test Results
Melting point | 198-201°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...