1-(2-Chlorophenylsulfonyl)-2-pyrrolidinecarboxylic acid ≥95%

Supplier: Matrix Scientific

042568-500MG 042568-1G 042568-5G
101856-398EA 181.54 USD
101856-398 101856-400 101856-402
1-(2-Chlorophenylsulfonyl)-2-pyrrolidinecarboxylic acid ≥95%
1-(2-Chlorophenylsulphonyl)-2-pyrrolidinecarboxylic acid

Hazard: Irritant

Formula: C₁₁H₁₂ClNO₄S
Melting Pt: 140…142 °C
MDL Number: MFCD00172807
CAS Number: 251096-95-4

Order Now

Specification Test Results

Melting point 140-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR