3-[(2-Methoxy-2-oxoethyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%

Supplier: Matrix Scientific

042566-500MG 042566-1G
101856-394EA 243.08 USD
101856-394 101856-396
3-[(2-Methoxy-2-oxoethyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%
3-[(2-Methoxy-2-oxoethyl)sulfanyl]-2-thiophenecarboxylic acid

Hazard: Irritant

Formula: C₈H₈O₄S₂
Melting Pt: 155…157 °C
MDL Number: MFCD00663874
CAS Number: 251096-93-2

Order Now

Specification Test Results

Melting point 155-157°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR