2-{[(4-Methoxyphenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%

Supplier: Matrix Scientific

042565-500MG 042565-1G 042565-5G
101856-388EA 278.46 USD
101856-388 101856-390 101856-392
2-{[(4-Methoxyphenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%
2-{[(4-Methoxyphenyl)sulfonyl]amino}-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₆H₁₇NO₅S
Melting Pt: 149…151 °C
MDL Number: MFCD00173013
CAS Number: 40280-00-0

Order Now

Specification Test Results

Melting point 149-151°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR