2-{[(4-Bromophenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%

Supplier: Matrix Scientific

042564-500MG 042564-5G
101856-382EA 181.54 USD
101856-382 101856-386
2-{[(4-Bromophenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%
2-{[(4-Bromophenyl)sulfonyl]amino}-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₅H₁₄BrNO₄S
Melting Pt: 165…167 °C
MDL Number: MFCD00173012
CAS Number: 37642-61-8

Order Now

Specification Test Results

Melting point 165-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR