1-Methyl-4-(4-nitrophenyl)piperazine ≥95%

Supplier: Matrix Scientific

042561-500MG 042561-1G 042561-5G
101856-366EA 181.54 USD
101856-366 101856-368 101856-370
1-Methyl-4-(4-nitrophenyl)piperazine ≥95%
1-Methyl-4-(4-nitrophenyl)piperazine

Hazard: Irritant

Formula: C₁₁H₁₅N₃O₂
MW: 221.26 g/mol
Melting Pt: 109…111 °C
MDL Number: MFCD00101003
CAS Number: 16155-03-6

Order Now

Specification Test Results

Melting point 109-111°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR