3-Methyl-2-({[3-(trifluoromethyl)phenyl]sulfonyl}amino)butanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Methyl-2-[3-(trifluoromethyl)benzenesulphonylamino]butyric acid

042560-500MG 042560-1G 042560-5G
101856-360EA 375.38 USD
101856-360 101856-362 101856-364
3-Methyl-2-({[3-(trifluoromethyl)phenyl]sulfonyl}amino)butanoic acid ≥95%
3-Methyl-2-({[3-(trifluoromethyl)phenyl]sulfonyl}amino)butanoic acid

Hazard: Irritant

Formula: C₁₂H₁₄F₃NO₄S
MW: 325.31 g/mol
Melting Pt: 127…129 °C
MDL Number: MFCD00173010
CAS Number: 250714-85-3

Order Now

Specification Test Results

Melting point 127-129°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR