3-Chloro-N-(4-morpholinophenyl)propanamide ≥95%

Supplier: Matrix Scientific

042559-500MG 042559-1G 042559-5G
101856-354EA 375.38 USD
101856-354 101856-356 101856-358
3-Chloro-N-(4-morpholinophenyl)propanamide ≥95%
3-Chloro-N-(4-morpholinophenyl)propanamide

Hazard: Irritant

Formula: C₁₃H₁₇ClN₂O₂
MW: 268.75 g/mol
Melting Pt: 180…182 °C
MDL Number: MFCD00172701
CAS Number: 250714-83-1

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR