2-{[(2-Chlorophenyl)sulfonyl]amino}-3-methylbutanoic acid ≥95%

Supplier: Matrix Scientific

042558-500MG 042558-1G 042558-5G
101856-348EA 375.38 USD
101856-348 101856-350 101856-352
2-{[(2-Chlorophenyl)sulfonyl]amino}-3-methylbutanoic acid ≥95%
2-{[(2-Chlorophenyl)sulfonyl]amino}-3-methylbutanoic acid

Hazard: Irritant

Formula: C₁₁H₁₄ClNO₄S
Melting Pt: 152…154 °C
MDL Number: MFCD00173009
CAS Number: 250714-82-0

Order Now

Specification Test Results

Melting point 152-154°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR