2-{[(4-Bromophenyl)sulfonyl]amino}-3-methylbutanoic acid ≥95%

Supplier: Matrix Scientific

042557-500MG 042557-1G 042557-5G
101856-342EA 181.54 USD
101856-342 101856-344 101856-346
2-{[(4-Bromophenyl)sulfonyl]amino}-3-methylbutanoic acid ≥95%
2-{[(4-Bromophenyl)sulfonyl]amino}-3-methylbutanoic acid

Hazard: Irritant

Formula: C₁₁H₁₄BrNO₄S
Melting Pt: 181…183 °C
MDL Number: MFCD00173008
CAS Number: 250714-81-9

Order Now

Specification Test Results

Melting point 181-183°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR