2-{[(4-Fluorophenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-Fluorobenzenesulphamido)-2-phenylacetic acid

042553-500MG 042553-1G 042553-5G
101856-324EA 181.54 USD
101856-324 101856-326 101856-328
2-{[(4-Fluorophenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%
2-{[(4-Fluorophenyl)sulfonyl]amino}-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₄H₁₂FNO₄S
MW: 309.32 g/mol
Melting Pt: 179…181 °C
MDL Number: MFCD00173006
CAS Number: 117309-49-6

Order Now

Specification Test Results

Melting point 179-181°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR