2-{[(4-Bromophenyl)sulfonyl]amino}propanoic acid ≥95%

Supplier: Matrix Scientific

042552-500MG 042552-1G 042552-5G
101856-318EA 181.54 USD
101856-318 101856-320 101856-322
2-{[(4-Bromophenyl)sulfonyl]amino}propanoic acid ≥95%
2-{[(4-Bromophenyl)sulfonyl]amino}propanoic acid

Hazard: Irritant

Formula: C₉H₁₀BrNO₄S
Melting Pt: 161…162 °C
MDL Number: MFCD00664349
CAS Number: 250714-65-9

Order Now

Specification Test Results

Melting point 161-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR