N-{[3-(Trifluoromethyl)phenyl]sulfonyl}-DL-phenylalanine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulphonyl}amino)propanoic acid

042551-500MG 042551-1G 042551-5G
101856-312EA 375.38 USD
101856-312 101856-314 101856-316
N-{[3-(Trifluoromethyl)phenyl]sulfonyl}-DL-phenylalanine ≥95%
N-{[3-(Trifluoromethyl)phenyl]sulfonyl}-DL-phenylalanine

Hazard: Irritant

Formula: C₁₆H₁₄F₃NO₄S
MW: 373.35 g/mol
Melting Pt: 140…142 °C
MDL Number: MFCD00173005
CAS Number: 250714-63-7

Order Now

Specification Test Results

Melting point 140-142°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR