2-{[(4-Chlorophenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%

Supplier: Matrix Scientific

042549-500MG 042549-1G 042549-5G
101856-300EA 278.46 USD
101856-300 101856-302 101856-304
2-{[(4-Chlorophenyl)sulfonyl]amino}-3-phenylpropanoic acid ≥95%
2-{[(4-Chlorophenyl)sulfonyl]amino}-3-phenylpropanoic acid

Hazard: Irritant

Formula: C₁₅H₁₄ClNO₄S
MW: 339.8 g/mol
Melting Pt: 147…149 °C
MDL Number: MFCD00173003
CAS Number: 40279-97-8

Order Now

Specification Test Results

Melting point 147-149°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR