2-({[3-(Trifluoromethyl)phenyl]sulfonyl}amino)propanoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[3-(Trifluoromethyl)benzenesulphonamido]propionic acid

042547-500MG 042547-1G 042547-5G
101856-294EA 375.38 USD
101856-294 101856-296 101856-298
2-({[3-(Trifluoromethyl)phenyl]sulfonyl}amino)propanoic acid ≥95%
2-({[3-(Trifluoromethyl)phenyl]sulfonyl}amino)propanoic acid

Hazard: Irritant

Formula: C₁₀H₁₀F₃NO₄S
MW: 297.25 g/mol
Melting Pt: 123…125 °C
MDL Number: MFCD00172805
CAS Number: 288266-54-6

Order Now

Specification Test Results

Melting point 123-125°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR