2-{[(4-Methylphenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

042546-500MG 042546-1G 042546-5G
101856-288EA 181.54 USD
101856-288 101856-290 101856-292
2-{[(4-Methylphenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%
2-{[(4-Methylphenyl)sulfonyl]amino}-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₄S
Melting Pt: 178…180 °C
MDL Number: MFCD00172804
CAS Number: 60712-47-2

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR