2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulfonyl}amino)acetic acid ≥95%

Supplier: Matrix Scientific

Synonyms: N-{[3-(Trifluoromethyl)phenyl]sulphonyl}-DL-phenylglycine

042544-500MG 042544-1G 042544-5G
101856-276EA 375.38 USD
101856-276 101856-278 101856-280
2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulfonyl}amino)acetic acid ≥95%
2-Phenyl-2-({[3-(trifluoromethyl)phenyl]sulfonyl}amino)acetic acid

Hazard: Irritant

Formula: C₁₅H₁₂F₃NO₄S
MW: 359.33 g/mol
Melting Pt: 169…171 °C
MDL Number: MFCD00172802
CAS Number: 250714-41-1

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR