2-{[(4-Methoxyphenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%

Supplier: Matrix Scientific

042543-500MG 042543-1G 042543-5G
101856-270EA 278.46 USD
101856-270 101856-272 101856-274
2-{[(4-Methoxyphenyl)sulfonyl]amino}-2-phenylacetic acid ≥95%
2-{[(4-Methoxyphenyl)sulfonyl]amino}-2-phenylacetic acid

Hazard: Irritant

Formula: C₁₅H₁₅NO₅S
Melting Pt: 171…173 °C
MDL Number: MFCD00172801
CAS Number: 117309-46-3

Order Now

Specification Test Results

Melting point 171-173°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR