3-[(3,4-Dichlorobenzyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
3-((3,4-Dichlorobenzyl)thio)thiophene-2-carboxylic acid
042542-500MG
042542-1G
101856-266EA
556.92
USD
101856-266
101856-268
3-[(3,4-Dichlorobenzyl)sulfanyl]-2-thiophenecarboxylic acid ≥95%
3-[(3,4-Dichlorobenzyl)sulfanyl]-2-thiophenecarboxylic acid
Hazard: Irritant
Formula:
C₁₂H₈Cl₂O₂S₂ MW: 319.23 g/mol Melting Pt: 189…190 °C |
MDL Number:
MFCD00663856 CAS Number: 251096-84-1 |
Specification Test Results
Melting point | 189-190°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...