Methyl-4-methyl-2-{[(4-methylphenyl)sulfonyl]amino}pentanoate ≥95%
Supplier: Matrix Scientific
042541-500MG
042541-1G
042541-5G
101856-260EA
181.54
USD
101856-260
101856-262
101856-264
Methyl-4-methyl-2-{[(4-methylphenyl)sulfonyl]amino}pentanoate ≥95%
Methyl-4-methyl-2-{[(4-methylphenyl)sulfonyl]amino}pentanoate
Hazard: Irritant
Formula:
C₁₄H₂₁NO₄S MW: 299.39 g/mol Melting Pt: 54…56.5 °C |
MDL Number:
MFCD00664346 CAS Number: 51220-84-9 |
Specification Test Results
Melting point | 54-56.5°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...