Methyl-2-{[(4-methylphenyl)sulfonyl]amino}-2-phenylacetate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-(4-methylphenylsulphonamido)-2-phenylacetate

042540-500MG 042540-5G
101856-254EA 181.54 USD
101856-254 101856-258
Methyl-2-{[(4-methylphenyl)sulfonyl]amino}-2-phenylacetate ≥95%
Methyl-2-{[(4-methylphenyl)sulfonyl]amino}-2-phenylacetate

Hazard: Irritant

Formula: C₁₆H₁₇NO₄S
MW: 319.38 g/mol
Melting Pt: 113…115 °C
MDL Number: MFCD00664343
CAS Number: 1233-56-3

Order Now

Specification Test Results

Melting point 113-115°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR